missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AHSP / ERAF Protein
A DNA sequence encoding the Homo sapiens (Human) AHSP / ERAF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8157-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001305150.1 | |
AHSP / ERAF Protein | |
Research Use Only | |
AHSP | |
Recombinant Protein | |
E. coli | |
MALLKANKDL ISAGLKEFSV LLNQQVFNDP LVSEEDMVTV VEDWMNFYIN YYRQQVTGEP QERDKALQEL RQELNTLANP FLAKYRDFLK SHELPSHPPP SS | |
Greater than 80% as determined by SDS-PAGE. |
51327 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |