missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6431-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_000897.3 | |
Atrial natriuretic peptide receptor 1 Protein | |
Research Use Only | |
NPR1 | |
Recombinant Protein | |
Yeast | |
GNLTVAVVLP LANTSYPWSW ARVGPAVELA LAQVKARPDL LPGWTVRTVL GSSENALGVC SDTAAPLAAV DLKWEHNPAV FLGPGCVYAA APVGRFTAHW RVPLLTAGAP ALGFGVKDEY ALTTRAGPSY AKLGDFVAAL HRRLGWERQA LMLYAYRPGD EEHCFFLVEG LFMRVRDRLN ITVDHLEFAE DDLSHYTRLL RTMPRKGRVI YICSSPDAFR TLMLLALEAG LCGEDYVFFH LDIFGQSLQG GQGPAPRRPW ERGDGQDVSA RQAFQAAKII TYKDPDNPEY LEFLKQLKHL AYEQFNFTME DGLVNTIPAS FHDGLLLYIQ AVTETLAHGG TVTDGENITQ RMWNRSFQGV TGYLKIDSSG DRETDFSLWD MDPENGAFRV VLNYNGTSQE LVAVSGRKLN WPLGYPPPDI PKCGFDNEDP ACNQDHLSTL E | |
Greater than 90% as determined by SDS-PAGE. |
4881 | |
200 ÎĽg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 1 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction