missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-8 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-8, was expressed in the hosts and tags indicated.
2325.00 SEK - 18390.00 SEK
Specifications
Name | Caspase-8 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CASP8 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067181
|
enQuireBio™
QP8921-EC-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087181
|
enQuireBio™
QP8921-EC-200UG |
200 μg |
9835.00 SEK
200µg |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057181
|
enQuireBio™
QP8921-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007191
|
enQuireBio™
QP8921-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097181
|
enQuireBio™
QP8921-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077181
|
enQuireBio™
QP8921-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Caspase-8 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
CASP8 | |
Recombinant Protein | |
E. coli | |
SESQTLDKVY QMKSKPRGYC LIINNHNFAK AREKVPKLHS IRDRNGTHLD AGALTTTFEE LHFEIKPHDD CTVEQIYEIL KIYQLMDHSN MDCFICCILS HGDKGIIYGT DGQEAPIYEL TSQFTGLKCP SLAGKPKVFF IQACQGDNYQ KGIPVETDSE EQPYLEMDLS SPQTRYIPDE ADFLLGMATV NNCVSYRNPA EGTWYIQSLC QSLRERCPRG DDILTILTEV NYEVSNKDDK KNMGKQMPQP TFTLRKKLVF PSD | |
Greater than 90% as determined by SDS-PAGE. |