missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHCHD4 Protein
A DNA sequence encoding the Homo sapiens (Human) CHCHD4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7968-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091972.1 | |
CHCHD4 Protein | |
Research Use Only | |
CHCHD4 | |
Recombinant Protein | |
E. coli | |
MSYCRQEGKD RIIFVTKEDH ETPSSAELVA DDPNDPYEEH GLILPNGNIN WNCPCLGGMA SGPCGEQFKS AFSCFHYSTE EIKGSDCVDQ FRAMQECMQK YPDLYPQEDE DEEEEREKKP AEQAEETAPI EATATKEEEG SS | |
Greater than 90% as determined by SDS-PAGE. |
131474 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |