missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHCHD4 Protein
A DNA sequence encoding the Homo sapiens (Human) CHCHD4, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001091972.1 |
---|---|
Gene ID (Entrez) | 131474 |
Name | CHCHD4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051891
|
enQuireBio™
QP7968-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071891
|
enQuireBio™
QP7968-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061891
|
enQuireBio™
QP7968-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081891
|
enQuireBio™
QP7968-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091891
|
enQuireBio™
QP7968-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001901
|
enQuireBio™
QP7968-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001091972.1 | |
CHCHD4 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
131474 | |
Research Use Only | |
CHCHD4 | |
Recombinant Protein | |
E. coli | |
MSYCRQEGKD RIIFVTKEDH ETPSSAELVA DDPNDPYEEH GLILPNGNIN WNCPCLGGMA SGPCGEQFKS AFSCFHYSTE EIKGSDCVDQ FRAMQECMQK YPDLYPQEDE DEEEEREKKP AEQAEETAPI EATATKEEEG SS | |
Greater than 90% as determined by SDS-PAGE. |