Learn More
enQuireBio™ Recombinant Human Coagulation Factor VII / FVII / F7 Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor VII / FVII / F7, was expressed in the hosts and tags indicated.
2515.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_000122.1 |
---|---|
Gene ID (Entrez) | 2155 |
Name | Coagulation Factor VII / FVII / F7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909359
|
enQuireBio™
QP6012-ec-10ug |
10 ÎĽg | N/A | ||||||
15949359
|
enQuireBio™
QP6012-ec-50ug |
50 ÎĽg | N/A | ||||||
15999349
|
enQuireBio™
QP6012-ec-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15929359
|
enQuireBio™
QP6012-ec-200ug |
200 ÎĽg | N/A | ||||||
15939359
|
enQuireBio™
QP6012-ec-500ug |
500 ÎĽg | N/A | ||||||
15919359
|
enQuireBio™
QP6012-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000122.1 | |
Coagulation Factor VII / FVII / F7 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2155 | |
Research Use Only | |
F7 | |
Recombinant Protein | |
E. coli | |
ANAFLEELRP GSLERECKEE QCSFEEAREI FKDAERTKLF WISYSDGDQC ASSPCQNGGS CKDQLQSYIC FCLPAFEGRN CETHKDDQLI CVNENGGCEQ YCSDHTGTKR SCRCHEGYSL LADGVSCTPT VEYPCGKIPI LEKRNASKPQ GR | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Coagulation Factor VII / FVII / F7 Protein