missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome c-type heme lyase Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome c-type heme lyase, was expressed in the hosts and tags indicated.

2430.00 SEK - 32715.00 SEK

Specifications

Name Cytochrome c-type heme lyase Protein
Product Type Recombinant Protein
Protein Tag GST
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16091170
View Documents Promotion Details
enQuireBio™
QP6149-EC-100UG
100 μg
7400.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 15-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16011180
View Documents Promotion Details
enQuireBio™
QP6149-EC-1MG
1 mg
32715.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 15-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16001180
View Documents Promotion Details
enQuireBio™
QP6149-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021180
View Documents Promotion Details
enQuireBio™
QP6149-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16031180
View Documents Promotion Details
enQuireBio™
QP6149-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16041180
View Documents Promotion Details
enQuireBio™
QP6149-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Cytochrome c-type heme lyase Protein
GST
Human
Greater than 80% as determined by SDS-PAGE.
NP_001116080.1
Recombinant Protein
E. coli
MGLSPSAPAV AVQASNASAS PPSGCPMHEG KMKGCPVNTE PSGPTCEKKT YSVPAHQERA YEYVECPIRG TAAENKENLD PSNLMPPPNQ TPAPDQPFAL STVREESSIP RADSEKKWVY PSEQMFWNAM LKKGWKWKDE DISQKDMYNI IRIHNQNNEQ AWKEILKWEA LHAAECPCGP SLIRFGGKAK EYSPRARIRS WMGYELPFDR HDWIINRCGT EVRYVIDYYD GGEVNKDYQF TILDVRPALD SLSAVWDRMK VAWWRWTS
3052
HCCS
SDS
Product Certifications