missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytosolic 5'-nucleotidase 1A Protein

A DNA sequence encoding the Homo sapiens (Human) Cytosolic 5'-nucleotidase 1A, was expressed in the hosts and tags indicated.

2005.00 SEK - 22130.00 SEK

Specifications

Accession Number NP_115915.1
Gene ID (Entrez) 84618
Name Cytosolic 5'-nucleotidase 1A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16078931
View Documents
enQuireBio™
QP9892-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16018941
View Documents
enQuireBio™
QP9892-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068931
View Documents
enQuireBio™
QP9892-ye-100ug
100 ÎĽg
7570.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16098931
View Documents
enQuireBio™
QP9892-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008941
View Documents
enQuireBio™
QP9892-ye-500ug
500 ÎĽg
20530.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16088931
View Documents
enQuireBio™
QP9892-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_115915.1
Cytosolic 5'-nucleotidase 1A Protein
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
84618
Research Use Only
NT5C1A
Recombinant Protein
Yeast
MEPGQPREPQ EPREPGPGAE TAAAPVWEEA KIFYDNLAPK KKPKSPKPQN AVTIAVSSRA LFRMDEEQQI YTEQGVEEYV RYQLEHENEP FSPGPAFPFV KALEAVNRRL RELYPDSEDV FDIVLMTNNH AQVGVRLINS INHYDLFIER FCMTGGNSPI CYLKAYHTNL YLSADAEKVR EAIDEGIAAA TIFSPSRDVV VSQSQLRVAF DGDAVLFSDE SERIVKAHGL DRFFEHEKAH ENKPLAQGPL KGFLEALGRL QKKFYSKGLR LECPIRTYLV TARSAASSGA RALKTLRSWG LETDEALFLA GAPKGPLLEK IRPHIFFDDQ MFHVAGAQEM GTVAAHVPYG VAQTPRRTAP AKQAPSAQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Cytosolic 5'-nucleotidase 1A Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.