missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCXR / HCR2 Protein
A DNA sequence encoding the Homo sapiens (Human) DCXR / HCR2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7792-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001182147.1 | |
DCXR / HCR2 Protein | |
Research Use Only | |
DCXR | |
Recombinant Protein | |
E. coli | |
MELFLAGRRV LVTGAGKGIG RGTVQALHAT GARVVAVSRT QADLDSLVRE CPGIEPVCVD LGDWEATERA LGSVGPVDLL VNNAAVALLQ PFLEVTKEAF DRSFEVNLRA VIQVSQIVAR GLIARGVPGA IVNVSSQCSQ RAVTNHSVYC STKGALDMLT KVMALELGPH KIRVNAVNPT VVMTSMGQAT WSDPHKAKTM LNRIPLGKFA EVEHVVNAIL FLLSDRSGMT TGSTLPVEGG FWAC | |
Greater than 80% as determined by SDS-PAGE. |
51181 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human DCXR / HCR2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction