missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dickkopf-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Dickkopf-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8081-ec-200ug

 View more versions of this product

Product Code. 16052551

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_055236.1
Dickkopf-related protein 1
Research Use Only
DKK2
Recombinant Protein
E. coli
KLNSIKSSLG GETPGQAANR SAGMYQGLAF GGSKKGKNLG QAYPCSSDKE CEVGRYCHSP HQGSSACMVC RRKKKRCHRD GMCCPSTRCN NGICIPVTES ILTPHIPALD GTRHRDRNHG HYSNHDLGWQ NLGRPHTKMS HIKGHEGDPC LRSSDCIEGF CCARHFWTKI CKPVLHQGEV CTKQRKKGSH GLEIFQRCDC AKGLSCKVWK DATYSSKARL HVCQKI
Greater than 90% as determined by SDS-PAGE.
27123
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Dickkopf-related protein 1 > 200μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.