missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA primase small subunit Protein

A DNA sequence encoding the Homo sapiens (Human) DNA primase small subunit, was expressed in the hosts and tags indicated.

1640.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_000937.1
Gene ID (Entrez) 5557
Name DNA primase small subunit Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16013950
View Documents
enQuireBio™
QP6528-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16053950
View Documents
enQuireBio™
QP6528-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16003950
View Documents
enQuireBio™
QP6528-ec-100ug
100 ÎĽg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16033950
View Documents
enQuireBio™
QP6528-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16043950
View Documents
enQuireBio™
QP6528-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16023950
View Documents
enQuireBio™
QP6528-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_000937.1
DNA primase small subunit Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5557
Research Use Only
PRIM1
Recombinant Protein
E. coli
METFDPTELP ELLKLYYRRL FPYSQYYRWL NYGGVIKNYF QHREFSFTLK DDIYIRYQSF NNQSDLEKEM QKMNPYKIDI GAVYSHRPNQ HNTVKLGAFQ AQEKELVFDI DMTDYDDVRR CCSSADICPK CWTLMTMAIR IIDRALKEDF GFKHRLWVYS GRRGVHCWVC DESVRKLSSA VRSGIVEYLS LVKGGQDVKK KVHLSEKIHP FIRKSINIIK KYFEEYALVN QDILENKESW DKILALVPET IHDELQQSFQ KSHNSLQRWE HLKKVASRYQ NNIKNDKYGP WLEWEIMLQY CFPRLDINVS KGINHLLKSP FSVHPKTGRI SVPIDLQKVD QFDPFTVPTI SFICRELDAI STNEEEKEEN EAESDVKHRT RDYKKTSLAP YVKVFEHFLE NLDKSRKGEL LKKSDLQKDF
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human DNA primase small subunit Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.