missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MYLIP Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MYLIP, was expressed in the hosts and tags indicated.

1580.00 SEK - 22920.00 SEK

Specifications

Name E3 ubiquitin-protein ligase MYLIP Protein
Product Type Recombinant Protein
Protein Tag GST
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16012051
View Documents Promotion Details
enQuireBio™
QP7993-EC-100UG
100 μg
5025.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 15-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16032051
View Documents Promotion Details
enQuireBio™
QP7993-EC-1MG
1 mg
22920.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 15-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16022051
View Documents Promotion Details
enQuireBio™
QP7993-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16042051
View Documents Promotion Details
enQuireBio™
QP7993-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16052051
View Documents Promotion Details
enQuireBio™
QP7993-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16062051
View Documents Promotion Details
enQuireBio™
QP7993-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

E3 ubiquitin-protein ligase MYLIP Protein
GST
Human
Greater than 90% as determined by SDS-PAGE.
NP_037394.2
Recombinant Protein
E. coli
MLCYVTRPDA VLMEVEVEAK ANGEDCLNQV CRRLGIIEVD YFGLQFTGSK GESLWLNLRN RISQQMDGLA PYRLKLRVKF FVEPHLILQE QTRHIFFLHI KEALLAGHLL CSPEQAVELS ALLAQTKFGD YNQNTAKYNY EELCAKELSS ATLNSIVAKH KELEGTSQAS AEYQVLQIVS AMENYGIEWH SVRDSEGQKL LIGVGPEGIS ICKDDFSPIN RIAYPVVQMA TQSGKNVYLT VTKESGNSIV LLFKMISTRA ASGLYRAITE THAFYRCDTV TSAVMMQYSR DLKGHLASLF LNENINLGKK YVFDIKRTSK EVYDHARRAL YNAGVVDLVS RNNQSPSHSP LKSSESSMNC SSCEGLSCQQ TRVLQEKLRK LKEAMLCMVC CEEEINSTFC PCGHTVCCES CAAQLQSCPV CRSRVEHVQH VYLPTHTSLL NLTVI
29116
MYLIP
SDS
Product Certifications