missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F-box / LRR-repeat protein 1

A DNA sequence encoding the Homo sapiens (Human) F-box/LRR-repeat protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8251-ec-500ug

 View more versions of this product

Product Code. 16063571

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001165184.1
F-box / LRR-repeat protein 1
Research Use Only
FBXL2
Recombinant Protein
E. coli
MVFSNNDEGL INKKLPKELL LRIFSFLDIV TLCRCAQISK AWNILALDGS NWQRIDLFNF QTDVEGRVVE NISKRCGGFL RKLSLRGCIG VGDSSLKTFA QNCRNIEHLN LNGCTKITDS TCYSLSRFCS KLKHLDLTSC VSITNSSLKG ISEGCRNLEY LNLSWCDQIT KDGIEALVRG CRGLKALLLR GCTQLEDEAL KHIQNYCHEL VSLNLQSCSR ITDEGVVQIC RGCHRLQALC LSGCSNLTDA SLTALGLNCP RLQILEAARC SHLTDAGFTL LARNCHELEK MDLEECILIT DSTLIQLSIH CPKLQALSLS HCELITDDGI LHLSNSTCGH ERLRVLELDN CLLITDVALE HLENCRGLER LELYDCQQVT RAGIKRMRAQ LPHVKVHAYF APVTPPTAVA GSGQRLCRCC VIL
Greater than 90% as determined by SDS-PAGE.
25827
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human F-box / LRR-repeat protein 1 > 500μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.