missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human GLP-1R / GLP1R Protein Product Code.: 16060740

enQuireBio™ Recombinant Human GLP-1R / GLP1R Protein

Product Code. 16060740
50 μg, 50µg
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 16060740

Brand: enQuireBio™ QP6090ma50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_002053.3
Gene ID (Entrez) 2740
Name GLP-1R / GLP1R Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Glp1r
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag His
Sequence RPQGATVSLW ETVQKWREYR RQCQRSLTED PPPATDLFCN RTFDEYACWP DGEPGSFVNV SCPWYLPWAS SVPQGHVYRF CTAEGLWLQK DNSSLPWRDL SECEESKRGE RSSPEEQLLF LY
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 98.9% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human GLP-1R / GLP1R Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.