missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glucosidase 2 subunit beta Protein

A DNA sequence encoding the Homo sapiens (Human) Glucosidase 2 subunit beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8431-ec-200ug

 View more versions of this product

Product Code. 16034581

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001001329.1
Glucosidase 2 subunit beta Protein
Research Use Only
PRKCSH
Recombinant Protein
E. coli
VEVKRPRGVS LTNHHFYDES KPFTCLDGSA TIPFDQVNDD YCDCKDGSDE PGTAACPNGS FHCTNTGYKP LYIPSNRVND GVCDCCDGTD EYNSGVICEN TCKEKGRKER ESLQQMAEVT REGFRLKKIL IEDWKKAREE KQKKLIELQA GKKSLEDQVE MLRTVKEEAE KPEREAKEQH QKLWEEQLAA AKAQQEQELA ADAFKELDDD MDGTVSVTEL QTHPELDTDG DGALSEAEAQ ALLSGDTQTD ATSFYDRVWA AIRDKYRSEA LPTDLPAPSA PDLTEPKE
Greater than 90% as determined by SDS-PAGE.
5589
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Glucosidase 2 subunit beta Protein > 200μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.