missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Glutaredoxin-3 Protein Product Code.: 16010770

enQuireBio™ Recombinant Human Glutaredoxin-3 Protein

Product Code. 16010770
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16010770

Brand: enQuireBio™ QP6094ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin-3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001186797.1
Gene ID (Entrez) 10539
Name Glutaredoxin-3 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GLRX3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence AAGAAEAAVA AVEEVGSAGQ FEELLRLKAK SLLVVHFWAP WAPQCAQMNE VMAELAKELP QVSFVKLEAE GVPEVSEKYE ISSVPTFLFF KNSQKIDRLD GAHAPELTKK VQRHASSGSF LPSANEHLKE DLNLRLKKLT HAAPCMLFMK GTPQEPRCGF SKQMVEILHK HNIQFSSFDI FSDEEVRQGL KAYSSWPTYP QLYVSGELIG GLDIIKELEA SEELDTICPK APKLEERLKV LTNKASVMLF MKGNKQEAKC GFSKQILEIL NSTGVEYETF DILEDEEVRQ GLKAYSNWPT YPQLYVKGEL VGGLDIVKEL KENGELLPIL RGEN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Glutaredoxin-3 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.