missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8472-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001244222.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
Research Use Only | |
HNRNPH1 | |
Recombinant Protein | |
E. coli | |
MLGTEGGEGF VVKVRGLPWS CSADEVQRFF SDCKIQNGAQ GIRFIYTREG RPSGEAFVEL ESEDEVKLAL KKDRETMGHR YVEVFKSNNV EMDWVLKHTG PNSPDTANDG FVRLRGLPFG CSKEEIVQFF SGLEIVPNGI TLPVDFQGRS TGEAFVQFAS QEIAEKALKK HKERIGHRYI EIFKSSRAEV RTHYDPPRKL MAMQRPGPYD RPGAG | |
Greater than 90% as determined by SDS-PAGE. |
3187 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |