missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
Accession Number | NP_057856.1 |
Gene ID (Entrez) | 155971 |
Name | HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein |
Quantity | 100 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | gp120 |
Protein Length | Full Length |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein >
Spot an opportunity for improvement?Share a Content Correction