missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-F Protein
A cDNA sequence encoding the HLA-F was constructed and used to recombinantly synthesize the protein.
1615.00 SEK - 20560.00 SEK
Specifications
Gene ID (Entrez) | 3134 |
---|---|
Name | HLA-F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HLA-F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919918
|
enQuireBio™
QP12277-2ug |
2 ÎĽg |
1615.00 SEK
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15909918
|
enQuireBio™
QP12277-10ug |
10 ÎĽg |
2515.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15999908
|
enQuireBio™
QP12277-100ug |
100 ÎĽg |
20560.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
3134 | |
Research Use Only | |
HLA-F | |
Human | |
His | |
HLA-F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl 20% glycerol and 1mM DTT. |
HLA-F Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGSHSLRY FSTAVSRPGR GEPRYIAVEY VDDTQFLRFD SDAAIPRMEP REPWVEQEGP QYWEWTTGYA KANAQTDRVA LRNLLRRYNQ SEAGSHTLQG MNGCDMGPDG RLLRGYHQHA YDGKDYISLN EDLRSWTAAD TVAQITQRFY EAEEYAEEFR TYLEGECLEL LRRYLENGKE TLQRADPPKA HVAHHPISDH EATLRCWALG FYPAEITLTW QRDGEEQTQD TELVETRPAG DGTFQKWAAV VVPPGEEQRY TCHVQHEGLP QPLILRWEQS PQPTIPI | |
Greater than 90.0% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human HLA-F Protein