missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HRASLS3 Protein
A cDNA sequence encoding the HRASLS3 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 30850.00 SEK
Specifications
Name | HRASLS3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940019
|
enQuireBio™
QP12312-10UG |
10 μg |
1545.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960019
|
enQuireBio™
QP12312-50UG |
50 μg |
2405.00 SEK
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950019
|
enQuireBio™
QP12312-1MG |
1 mg |
30850.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HRASLS3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein contains 20mM Tris-HCl buffer pH 8. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRAPIPEPKP GDLIEIFRPF YRHWAIYVGD GYVVHLAPPS EVAGAGAASV MSALTDKAIV KKELLYDVAG SDKYQVNNKH DDKYSPLPCS KIIQRAEELV GQEVLYKLTS ENCEHFVNEL RYGVARSDQV RDV | |
Greater than 95.0% as determined by SDS-PAGE. |