Learn More
R&D Systems™ Recombinant Human IL-28A/IFN-lambda 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1587-IL-025
3964.00 SEK valid until 2025-07-30
Use promo code "24078" to get your promotional price.
 View more versions of this product
Product Code. 18156210
  • 4955.00 SEK / 25µg
Ultimate Promo May
Use promo code 24078 at checkout
View Available Promotions
Description
The Recombinant Human IL-28A/IFN-lambda 2 Protein is derived from NS0. The Recombinant Human IL-28A/IFN-lambda 2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lyophilized from a 0.2ÎĽm filtered solution in PBS with BSA as a carrier protein.
20.8 kDa
25 ÎĽg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
282616
IL-28A/IFN-lambda 2
Mouse myeloma cell line,NS0-derived human IL-28A/IFN-lambda 2 protein, (N-terminus) Human IL-28A Val26-Val200 Accession &Num; NP_742150 DI 6-His tag (C-terminus)
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Certificates
Promotions
Promotions
  • R&D Systems , Tocris
    Empowering Research: The Right Reagents for Every Step
    Promo Code: 24078
    Expires2025-07-30
Product Content Correction
Product Title
R&D Systems™ Recombinant Human IL-28A/IFN-lambda 2 Protein > 25ug