missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kynurenine 3-monooxygenase Protein

A DNA sequence encoding the Homo sapiens (Human) Kynurenine 3-monooxygenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6271-ye-10ug

 View more versions of this product

Product Code. 16082090

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_003670.2
Kynurenine 3-monooxygenase Protein
Research Use Only
KMO
Recombinant Protein
Yeast
MDSSVIQRKK VAVIGGGLVG SLQACFLAKR NFQIDVYEAR EDTRVATFTR GRSINLALSH RGRQALKAVG LEDQIVSQGI PMRARMIHSL SGKKSAIPYG TKSQYILSVS RENLNKDLLT AAEKYPNVKM HFNHRLLKCN PEEGMITVLG SDKVPKDVTC DLIVGCDGAY STVRSHLMKK PRFDYSQQYI PHGYMELTIP PKNGDYAMEP NYLHIWPRNT FMMIALPNMN KSFTCTLFMP FEEFEKLLTS NDVVDFFQKY FPDAIPLIGE KLLVQDFFLL PAQPMISVKC SSFHFKSHCV LLGDAAHAIV PFFGQGMNAG FEDCLVFDEL MDKFSNDLSL CLPVFSRLRI PDDHAISDLS MYNYIEMRAH VNSSWFIFQK NMERFLHAIM PSTFIPLYTM VTFSRIRYHE AVQRWHWQKK VINKGLFFLG SLIAISSTYL LIHYMSPRSF LRLRRPWNWI AHFRNTTCFP AKAVDSLEQI SNLISR
Greater than 90% as determined by SDS-PAGE.
8564
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Kynurenine 3-monooxygenase Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.