missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human LGR5 Protein Product Code.: 16057911

enQuireBio™ Recombinant Human LGR5 Protein

Product Code. 16057911
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16057911

Brand: enQuireBio™ QP9247ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) LGR5, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001264155.1
Gene ID (Entrez) 8549
Name LGR5 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol LGR5
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence GSSPRSGVLL RGCPTHCHCE PDGRMLLRVD CSDLGLSELP SNLSVFTSYL DLSMNNISQL LPNPLPSLRF LEELRLAGNA LTYIPKGAFT GLYSLKVLML QNNQLRHVPT EALQNLRSLQ SLRLDANHIS YVPPSCFSGL HSLRHLWLDD NALTEIPVQA FRSLSALQAM TLALNKIHHI PDYAFGNLSS LVVLHLHNNR IHSLGKKCFD GLHSLETLDL NYNNLDEFPT AIRTLSNLKE LGFHSNNIRS IPEKAFVGNP SLITIHFYDN PIQFVGRSAF QHLPELRTLT LNGASQITEF PDLTGTANLE SLTLTGAQIS SLPQTVCNQL PNLQVLDLSY NLLEDLPSFS VCQKLQKIDL RHNEIYEIKV DTFQQLLSLR SLNLAWNKIA IIHPNAFSTL PSLIKLDLSS NLLSSFPITG LHGLTHLKLT GNHALQSLIS SENFPELKVI EMPYAYQCCA FGVCENAYKI SNQWNKGDNS SMDDLHKKDA GMFQAQDERD LEDFLLDFEE DLKALHSVQC SPSPGPFKPC EHLLDGWLIR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human LGR5 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.