missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Ly6/PLAUR domain-containing protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7780-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001182614.1 | |
Ly6 / PLAUR domain-containing protein 1 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
130574 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LYPD6 | |
Recombinant Protein | |
Yeast | |
AQSRDFTVKD IIYLHPSTTP YPGGFKCFTC EKAADNYECN RWAPDIYCPR ETRYCYTQHT MEVTGNSISV TKRCVPLEEC LSTGCRDSEH EGHKVCTSCC EGNICNLPLP RNETDATFAT TSPINQTNGH PRCMSVIVSC LWLWLGLML |