missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein Product Code.: 16042650

enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein

Product Code. 16042650
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16042650

Brand: enQuireBio™ QP6347ye1mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Mannan-binding lectin serine protease 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001027019.1
Gene ID (Entrez) 5648
Name Mannan-binding lectin serine protease 1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol MASP1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence HTVELNNMFG QIQSPGYPDS YPSDSEVTWN ITVPDGFRIK LYFMHFNLES SYLCEYDYVK VETEDQVLAT FCGRETTDTE QTPGQEVVLS PGSFMSITFR SDFSNEERFT GFDAHYMAVD VDECKEREDE ELSCDHYCHN YIGGYYCSCR FGYILHTDNR TCRVECSDNL FTQRTGVITS PDFPNPYPKS SECLYTIELE EGFMVNLQFE DIFDIEDHPE VPCPYDYIKI KVGPKVLGPF CGEKAPEPIS TQSHSVLILF HSDNSGENRG WRLSYRAAGN ECPELQPPVH GKIEPSQAKY FFKDQVLVSC DTGYKVLKDN VEMDTFQIEC LKDGTWSNKI PTCKIVDCRA PGELEHGLIT FSTRNNLTTY KSEIKYSCQE PYYKMLNNNT GIYTCSAQGV WMNKVLGRSL PTCLPVCGLP KFSRKLMARI FNGRPAQKGT TPWIAMLSHL NGQPFCGGSL LGSSWIVTAA HCLHQSLDPE DPTLRDSDLL SPSDFKIILG KHWRLRSDEN EQHLGVKHTT LHPQYDPNTF ENDVALVELL ESPVLNAFVM PICLPEGPQQ EGAMVIVSGW GKQFLQRFPE TLMEIEIPIV DHSTCQKAYA PLKKKVTRDM ICAGEKEGGK DACAGDSGGP MVTLNRERGQ WYLVGTVSWG DDCGKKDRYG VYSYIHHNKD WIQRVTGVRN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.