missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Molybdopterin synthase catalytic subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Molybdopterin synthase catalytic subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6373-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_004522.1 | |
Molybdopterin synthase catalytic subunit Protein | |
Research Use Only | |
MOCS2 | |
Recombinant Protein | |
E. coli | |
MSSLEISSSC FSLETKLPLS PPLVEDSAFE PSRKDMDEVE EKSKDVINFT AEKLSVDEVS QLVISPLCGA ISLFVGTTRN NFEGKKVISL EYEAYLPMAE NEVRKICSDI RQKWPVKHIA VFHRLGLVPV SEASIIIAVS SAHRAASLEA VSYAIDTLKA KVPIWKKEIY EESSTWKGNK ECFWASNS | |
Greater than 80% as determined by SDS-PAGE. |
4338 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Molybdopterin synthase catalytic subunit Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction