Learn More
enQuireBio™ Recombinant Human Myelin expression factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Myelin expression factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6399-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001288139.1 | |
Myelin expression factor 2 Protein | |
Research Use Only | |
MYEF2 | |
Recombinant Protein | |
E. coli | |
GGMNRIGGGI GFGGLEAMNS MGGFGGVGRM GELYRGAMTS SMERDFGRGD IGINQGFGDS FGRLGSAMIG GFAGRIGSSN MGPVGSGISG GMGSMNSVTG GMGMGLDRMS SSFDRMGPGI GAILERSIDM DRGFLSGPMG SGMRERIGSK GNQIFVRNLP FDLTWQKLKE KFSQCGHVMF AEIKMENGKS KGCGTVRFDS PESAE | |
Greater than 90% as determined by SDS-PAGE. |
50804 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Myelin expression factor 2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction