missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myoglobin His Protein
A cDNA sequence encoding the Myoglobin His was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12777-5ug
Additional Details : Weight : 0.01000kg
Specifications
4151 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MB | |
Human | |
His | |
The MB protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT, 100mM NaCl and 20% glycerol. |
Myoglobin His Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG |