missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin light chain 1 / 3, skeletal muscle isoform Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 1/3, skeletal muscle isoform, was expressed in the hosts and tags indicated.

1580.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_524144.1
Gene ID (Entrez) 4632
Name Myosin light chain 1 / 3, skeletal muscle isoform Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16093040
View Documents
enQuireBio™
QP6400-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16033050
View Documents
enQuireBio™
QP6400-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16083040
View Documents
enQuireBio™
QP6400-ec-100ug
100 ÎĽg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16013050
View Documents
enQuireBio™
QP6400-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16023050
View Documents
enQuireBio™
QP6400-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16003050
View Documents
enQuireBio™
QP6400-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_524144.1
Myosin light chain 1 / 3, skeletal muscle isoform Protein
Not determined.
Full Length of Isoform MLC3
Human
GST
Tris-based buffer, 50% glycerol
4632
Research Use Only
MYL1
Recombinant Protein
E. coli
MSFSADQIAE FKEAFLLFDR TGDSKITLSQ VGDVLRALGT NPTNAEVRKV LGNPSNEELN AKKIEFEQFL PMMQAISNNK DQATYEDFVE GLRVFDKEGN GTVMGAELRH VLATLGEKMK EEEVEALMAG QEDSNGCINY EAFVKHIMSI
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Myosin light chain 1 / 3, skeletal muscle isoform Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.