missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurocalcin-delta Protein
A DNA sequence encoding the Homo sapiens (Human) Neurocalcin-delta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6406-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001035714.1 | |
Neurocalcin-delta Protein | |
Research Use Only | |
NCALD | |
Recombinant Protein | |
E. coli | |
GKQNSKLRPE VMQDLLESTD FTEHEIQEWY KGFLRDCPSG HLSMEEFKKI YGNFFPYGDA SKFAEHVFRT FDANGDGTID FREFIIALSV TSRGKLEQKL KWAFSMYDLD GNGYISKAEM LEIVQAIYKM VSSVMKMPED ESTPEKRTEK IFRQMDTNRD GKLSLEEFIR GAKSDPSIVR LLQCDPSSAG QF | |
Greater than 80% as determined by SDS-PAGE. |
83988 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Neurocalcin-delta Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction