missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neuroglobin Protein
A DNA sequence encoding the Homo sapiens (Human) Neuroglobin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6418-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_067080.1 | |
Neuroglobin Protein | |
Research Use Only | |
NGB | |
Recombinant Protein | |
E. coli | |
MERPEPELIR QSWRAVSRSP LEHGTVLFAR LFALEPDLLP LFQYNCRQFS SPEDCLSSPE FLDHIRKVML VIDAAVTNVE DLSSLEEYLA SLGRKHRAVG VKLSSFSTVG ESLLYMLEKC LGPAFTPATR AAWSQLYGAV VQAMSRGWDG E | |
Greater than 90% as determined by SDS-PAGE. |
58157 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Neuroglobin Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction