missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear factor NF-kappa-B p105 subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear factor NF-kappa-B p105 subunit, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001158884.1 |
---|---|
Gene ID (Entrez) | 4790 |
Name | Nuclear factor NF-kappa-B p105 subunit Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045961
|
enQuireBio™
QP8688-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065961
|
enQuireBio™
QP8688-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055961
|
enQuireBio™
QP8688-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075961
|
enQuireBio™
QP8688-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085961
|
enQuireBio™
QP8688-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095961
|
enQuireBio™
QP8688-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001158884.1 | |
Nuclear factor NF-kappa-B p105 subunit Protein | |
Not determined. | |
Full length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4790 | |
Research Use Only | |
NFKB1 | |
Recombinant Protein | |
E. coli | |
MAEDDPYLGR PEQMFHLDPS LTHTIFNPEV FQPQMALPTD GPYLQILEQP KQRGFRFRYV CEGPSHGGLP GASSEKNKKS YPQVKICNYV GPAKVIVQLV TNGKNIHLHA HSLVGKHCED GICTVTAGPK DMVVGFANLG ILHVTKKKVF ETLEARMTEA CIRGYNPGLL VHPDLAYLQA EGGGDRQLGD REKELIRQAA LQQTKEMDLS VVRLMFTAFL PDSTGSFTRR LEPVVSDAIY DSKAPNASNL KIVRMDRTAG CVTGGEEIYL LCDKVQKDDI QIRFYEEEEN GGVWEGFGDF SPTDVHRQFA IVFKTPKYKD INITKPASVF VQLRRKSDLE TSEPKPFLYY PEIKDKEEVQ RKRQKLMPNF SDSFGGGSGA GAGGGGMFGS GGGGGGTGST GPGYSFPHYG FPTYGGITFH PGTTKSNAGM KHGTMDTESK KDPEGCDKSD DKNTVNLFGK VIETTEQDQE PSEATVGNGE VTLTYATGTK EESAGVQDNL FLEKAMQLAK RHANALFDYA VTGDVKMLLA VQRHLTAVQD ENGDSVLHLA IIHLHSQLVR DLLEVTSGLI SDDIINMRND LYQTPLHLAV ITKQEDVVED LLRAGADLSL LDRLGNSVLH LAAKEGHDKV LSILLKHKKA ALLLDHPNGD GLNAIHLAMM SNSLPCLLLL VAAGADVNAQ EQKSGRTALH LAVEHDNISL AGCLLLEGDA HVDSTTYDGT TPLHIAAGRG STRLAALLKA AGADPLVENF EPLYDLDDSW ENAGEDEGVV PGTTPLDMAT SWQVFDILNG KPYEPEFTSD DLLAQGDMKQ LAEDVKLQLY KLLEIPDPDK NWATLAQKLG LGILNNAFRL SPAPSKTLMD NYEVSGGTVR ELVEALRQMG YTEAIEVIQA ASSPVKTTSQ AHSLPLSPAS | |
Greater than 90% as determined by SDS-PAGE. |