missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Origin recognition complex subunit 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6453-ec-200ug

 View more versions of this product

Product Code. 16033420

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001177808.1
Origin recognition complex subunit 4 Protein
Research Use Only
ORC4
Recombinant Protein
E. coli
MSSRKSKSNS LIHTECLSQV QRILRERFCR QSPHSNLFGV QVQYKHLSEL LKRTALHGES NSVLIIGPRG SGKTMLINHA LKELMEIEEV SENVLQVHLN GLLQINDKIA LKEITRQLNL ENVVGDKVFG SFAENLSFLL EALKKGDRTS SCPVIFILDE FDLFAHHKNQ TLLYNLFDIS QSAQTPIAVI GLTCRLDILE LLEKRVKSRF SHRQIHLMNS FGFPQYVKIF KEQLSLPAEF PDKVFAEKWN ENVQYLSEDR SVQEVLQKHF NISKNLRSLH MLLMLALNRV TASHPFMTAV DLMEASQLCS MDSKANIVHG LSVLEICLII AMKHLNDIYE EEPFNFQMVY NEFQKFVQRK AHSVYNFEKP VVMKAFEHLQ QLELIKPMER TSGNSQREYQ LMKLLLDNTQ IMNALQKYPN CPTDVRQWAT SSLSWL
Greater than 90% as determined by SDS-PAGE.
5000
200 ÎĽg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Origin recognition complex subunit 4 Protein > 200ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.