missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Origin recognition complex subunit 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6453-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001177808.1 | |
Origin recognition complex subunit 4 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5000 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ORC4 | |
Recombinant Protein | |
E. coli | |
MSSRKSKSNS LIHTECLSQV QRILRERFCR QSPHSNLFGV QVQYKHLSEL LKRTALHGES NSVLIIGPRG SGKTMLINHA LKELMEIEEV SENVLQVHLN GLLQINDKIA LKEITRQLNL ENVVGDKVFG SFAENLSFLL EALKKGDRTS SCPVIFILDE FDLFAHHKNQ TLLYNLFDIS QSAQTPIAVI GLTCRLDILE LLEKRVKSRF SHRQIHLMNS FGFPQYVKIF KEQLSLPAEF PDKVFAEKWN ENVQYLSEDR SVQEVLQKHF NISKNLRSLH MLLMLALNRV TASHPFMTAV DLMEASQLCS MDSKANIVHG LSVLEICLII AMKHLNDIYE EEPFNFQMVY NEFQKFVQRK AHSVYNFEKP VVMKAFEHLQ QLELIKPMER TSGNSQREYQ LMKLLLDNTQ IMNALQKYPN CPTDVRQWAT SSLSWL |