missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POU domain class 2-associating factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) POU domain class 2-associating factor 1, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_006226.2 |
---|---|
Gene ID (Entrez) | 5450 |
Name | POU domain class 2-associating factor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019520
|
enQuireBio™
QP7577-ec-10ug |
10 μg | N/A | ||||||
16059520
|
enQuireBio™
QP7577-ec-50ug |
50 μg | N/A | ||||||
16009520
|
enQuireBio™
QP7577-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16039520
|
enQuireBio™
QP7577-ec-200ug |
200 μg | N/A | ||||||
16049520
|
enQuireBio™
QP7577-ec-500ug |
500 μg | N/A | ||||||
16029520
|
enQuireBio™
QP7577-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_006226.2 | |
POU domain class 2-associating factor 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5450 | |
Research Use Only | |
POU2AF1 | |
Recombinant Protein | |
E. coli | |
MLWQKPTAPE QAPAPARPYQ GVRVKEPVKE LLRRKRGHAS SGAAPAPTAV VLPHQPLATY TTVGPSCLDM EGSVSAVTEE AALCAGWLSQ PTPATLQPLA PWTPYTEYVP HEAVSCPYSA DMYVQPVCPS YTVVGPSSVL TYASPPLITN VTTRSSATPA VGPPLEGPEH QAPLTYFPWP QPLSTLPTST LQYQPPAPAL PGPQFVQLPI SIPEPVLQDM EDPRRAASSL TIDKLLLEEE DSDAYALNHT LSVEGF | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human POU domain class 2-associating factor 1 Protein