Learn More
enQuireBio™ Recombinant Human Protein E7 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7367-ye-100ug
 View more versions of this product
Product Code. 16088480
  • 7570.00 SEK / 100µg
Specifications
Protein E7 Protein
Research Use Only
E7
Recombinant Protein
Yeast
MHGPKATLQD IVLHLEPQNE IPVDLLCHEQ LSDSEEENDE IDGVNHQHLP ARRAEPQRHT MLCMCCKCEA RIKLVVESSA DDLRAFQQLF LNTLSFVCPW CASQQ
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Protein E7 Protein > 100μg