missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein ERGIC-53-like Protein
A DNA sequence encoding the Homo sapiens (Human) Protein ERGIC-53-like, was expressed in the hosts and tags indicated.
2075.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_068591.2 |
---|---|
Gene ID (Entrez) | 79748 |
Name | Protein ERGIC-53-like Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082401
|
enQuireBio™
QP8055-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022411
|
enQuireBio™
QP8055-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092401
|
enQuireBio™
QP8055-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012411
|
enQuireBio™
QP8055-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032411
|
enQuireBio™
QP8055-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002411
|
enQuireBio™
QP8055-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068591.2 | |
Protein ERGIC-53-like Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
79748 | |
Research Use Only | |
LMAN1L | |
Recombinant Protein | |
Yeast | |
GCPPLRRFEY KLSFKGPRLA LPGAGIPFWS HHGDAILGLE EVRLTPSMRN RSGAVWSRAS VPFSAWEVEV QMRVTGLGRR GAQGMAVWYT RGRGHVGSVL GGLASWDGIG IFFDSPAEDT QDSPAIRVLA SDGHIPSEQP GDGASQGLGS CHWDFRNRPH PFRARITYWG QRLRMSLNSG LTPSDPGEFC VDVGPLLLVP GGFFGVSAAT GTLADDHDVL SFLTFSLSEP SPEVPPQPFL EMQQLRLARQ LEGLWARLGL GTREDVTPKS DSEAQGEGER LFDLEETLGR HRRILQALRG LSKQLAQAER QWKKQLGPPG QARPDGGWAL DASCQIPSTP GRGGHLSMSL NKDSAKVGAL LHGQWTLLQA LQEMRDAAVR MAAEAQVSYL PVGIEHHFLE LDHILGLLQE ELRGPAKAAA KAPRPPGQPP RASSCLQ | |
Greater than 90% as determined by SDS-PAGE. |