Learn More
enQuireBio™ Recombinant Human Protein transport protein Sec16A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein transport protein Sec16A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6660-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001263347.1 | |
Protein transport protein Sec16A Protein | |
Research Use Only | |
SEC16A | |
Recombinant Protein | |
E. coli | |
AYLPDDKNKS IVWDEKKNQW VNLNEPEEEK KAPPPPPTSM PKTVQAAPPA LPGPPGAPVN MYSRRAAGTR ARYVDVLNPS GTQRSEPALA PADFVAPLAP LPIPSNLFVP TPDAEEPQLP DGTGREGPAA ARGLANPEPA PEPKVLSSAA SLPGSELPSS RPEGSQGGEL SRCSSMSSLS REVSQHFNQA PGDLPAAGGP PSGAMPFYNP AQ | |
Greater than 90% as determined by SDS-PAGE. |
9919 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |