missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein transport protein Sec16A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein transport protein Sec16A, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001263347.1 |
---|---|
Gene ID (Entrez) | 9919 |
Name | Protein transport protein Sec16A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064880
|
enQuireBio™
QP6660-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084880
|
enQuireBio™
QP6660-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074880
|
enQuireBio™
QP6660-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094880
|
enQuireBio™
QP6660-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004890
|
enQuireBio™
QP6660-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014890
|
enQuireBio™
QP6660-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001263347.1 | |
Protein transport protein Sec16A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
9919 | |
Research Use Only | |
SEC16A | |
Recombinant Protein | |
E. coli | |
AYLPDDKNKS IVWDEKKNQW VNLNEPEEEK KAPPPPPTSM PKTVQAAPPA LPGPPGAPVN MYSRRAAGTR ARYVDVLNPS GTQRSEPALA PADFVAPLAP LPIPSNLFVP TPDAEEPQLP DGTGREGPAA ARGLANPEPA PEPKVLSSAA SLPGSELPSS RPEGSQGGEL SRCSSMSSLS REVSQHFNQA PGDLPAAGGP PSGAMPFYNP AQ | |
Greater than 90% as determined by SDS-PAGE. |