Learn More
enQuireBio™ Recombinant Human Pterin-4-alpha-carbinolamine dehydratase Protein
A DNA sequence encoding the Homo sapiens (Human) Pterin-4-alpha-carbinolamine dehydratase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9320-ye-10ug
 View more versions of this product
Product Code. 16088091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_000272.1
Pterin-4-alpha-carbinolamine dehydratase Protein
Research Use Only
PCBD1
Recombinant Protein
Yeast
AGKAHRLSAE ERDQLLPNLR AVGWNELEGR DAIFKQFHFK DFNRAFGFMT RVALQAEKLD HHPEWFNVYN KVHITLSTHE CAGLSERDIN LASFIEQVAV SMT
Greater than 90% as determined by SDS-PAGE.
5092
10 ÎĽg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Pterin-4-alpha-carbinolamine dehydratase Protein > 10ÎĽg