missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human QPCTL Protein
A DNA sequence encoding the Homo sapiens (Human) QPCTL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8096-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001156849.1 | |
QPCTL Protein | |
Research Use Only | |
QPCTL | |
Recombinant Protein | |
E. coli | |
AAPVTLQLLF LDGEEALKEW GPKDSLYGSR HLAQLMESIP HSPGPTRIQA IELFMLLDLL GAPNPTFYSH FPRTVRWFHR LRSIEKRLHR LNLLQSHPQE VMYFQPGEPF GSVEDDHIPF LRRGVPVLHL ISTPFPAVWH TPADTEVNLH PPTVHNLCRI LAVFLAEYLG L | |
Greater than 90% as determined by SDS-PAGE. |
54814 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |