missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1 Product Code.: 16031341

enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1

Product Code. 16031341
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16031341

Brand: enQuireBio™ QP7878ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Regulator of microtubule dynamics protein 3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001291731.1
Gene ID (Entrez) 55177
Name Regulator of microtubule dynamics protein 1
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RMDN3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MSRLGALGGA RAGLGLLLGT AAGLGFLCLL YSQRWKRTQR HGRSQSLPNS LDYTQTSDPG RHVMLLRAVP GGAGDASVLP SLPREGQEKV LDRLDFVLTS LVALRREVEE LRSSLRGLAG EIVGEVRCHM EENQRVARRR RFPFVRERSD STGSSSVYFT ASSGATFTDA ESEGGYTTAN AESDNERDSD KESEDGEDEV SCETVKMGRK DSLDLEEEAA SGASSALEAG GSSGLEDVLP LLQQADELHR GDEQGKREGF QLLLNNKLVY GSRQDFLWRL ARAYSDMCEL TEEVSEKKSY ALDGKEEAEA ALEKGDESAD CHLWYAVLCG QLAEHESIQR RIQSGFSFKE HVDKAIALQP ENPMAHFLLG RWCYQVSHLS WLEKKTATAL LESPLSATVE DALQSFLKAE ELQPGFSKAG RVYISKCYRE LGKNSEARWW MKLALELPDV TKEDLAIQKD LEELEVILRD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.