missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RIPK2 Protein

A DNA sequence encoding the Homo sapiens (Human) RIPK2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6609-ec-10ug

 View more versions of this product

Product Code. 16014520

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_003812.1
RIPK2 Protein
Research Use Only
RIPK2
Recombinant Protein
E. coli
MNGEAICSAL PTIPYHKLAD LRYLSRGASG TVSSARHADW RVQVAVKHLH IHTPLLDSER KDVLREAEIL HKARFSYILP ILGICNEPEF LGIVTEYMPN GSLNELLHRK TEYPDVAWPL RFRILHEIAL GVNYLHNMTP PLLHHDLKTQ NILLDNEFHV KIADFGLSKW RMMSLSQSRS SKSAPEGGTI IYMPPENYEP GQKSRASIKH DIYSYAVITW EVLSRKQPFE DVTNPLQIMY SVSQGHRPVI NEESLPYDIP HRARMISLIE SGWAQNPDER PSFLKCLIEL EPVLRTFEEI TFLEAVIQLK KTKLQSVSSA IHLCDKKKME LSLNIPVNHG PQEESCGSSQ LHENSGSPET SRSLPAPQDN DFLSRKAQDC YFMKLHHCPG NHSWDSTISG SQRAAFCDHK TTPCSSAIIN PLSTAGNSER LQPGIAQQWI QSKREDIVNQ MTEACLNQSL DALLSRDLIM KEDYELVSTK PTRTSKVRQL LDTTDIQGEE FAKVIVQKLK DNKQMGLQPY PEILVVSRSP SLNLLQNKSM
Greater than 90% as determined by SDS-PAGE.
8767
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human RIPK2 Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.