Learn More
enQuireBio™ Recombinant Human RIPK2 Protein
A DNA sequence encoding the Homo sapiens (Human) RIPK2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6609-ec-50ug
 View more versions of this product
Product Code. 16054520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_003812.1
RIPK2 Protein
Research Use Only
RIPK2
Recombinant Protein
E. coli
MNGEAICSAL PTIPYHKLAD LRYLSRGASG TVSSARHADW RVQVAVKHLH IHTPLLDSER KDVLREAEIL HKARFSYILP ILGICNEPEF LGIVTEYMPN GSLNELLHRK TEYPDVAWPL RFRILHEIAL GVNYLHNMTP PLLHHDLKTQ NILLDNEFHV KIADFGLSKW RMMSLSQSRS SKSAPEGGTI IYMPPENYEP GQKSRASIKH DIYSYAVITW EVLSRKQPFE DVTNPLQIMY SVSQGHRPVI NEESLPYDIP HRARMISLIE SGWAQNPDER PSFLKCLIEL EPVLRTFEEI TFLEAVIQLK KTKLQSVSSA IHLCDKKKME LSLNIPVNHG PQEESCGSSQ LHENSGSPET SRSLPAPQDN DFLSRKAQDC YFMKLHHCPG NHSWDSTISG SQRAAFCDHK TTPCSSAIIN PLSTAGNSER LQPGIAQQWI QSKREDIVNQ MTEACLNQSL DALLSRDLIM KEDYELVSTK PTRTSKVRQL LDTTDIQGEE FAKVIVQKLK DNKQMGLQPY PEILVVSRSP SLNLLQNKSM
Greater than 90% as determined by SDS-PAGE.
8767
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Human RIPK2 Protein > 50μg