missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A12 / CAGC / Calgranulin-C Protein
A DNA sequence encoding the Homo sapiens (Human) S100A12 / CAGC / Calgranulin-C Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6637-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_005612.1 | |
S100A12 / CAGC / Calgranulin-C Protein | |
Research Use Only | |
S100A12 | |
Recombinant Protein | |
E. coli | |
TKLEEHLEGI VNIFHQYSVR KGHFDTLSKG ELKQLLTKEL ANTIKNIKDK AVIDEIFQGL DANQDEQVDF QEFISLVAIA LKAAHYHTHK E | |
Greater than 80% as determined by SDS-PAGE. |
6283 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human S100A12 / CAGC / Calgranulin-C Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction