missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Secreted Ly-6 / uPAR-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Secreted Ly-6/uPAR-related protein 1, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_065160.1 |
---|---|
Gene ID (Entrez) | 57152 |
Name | Secreted Ly-6 / uPAR-related protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16005190
|
enQuireBio™
QP6696-ec-10ug |
10 ÎĽg | N/A | ||||||
16045190
|
enQuireBio™
QP6696-ec-50ug |
50 ÎĽg | N/A | ||||||
16095180
|
enQuireBio™
QP6696-ec-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16025190
|
enQuireBio™
QP6696-ec-200ug |
200 ÎĽg | N/A | ||||||
16035190
|
enQuireBio™
QP6696-ec-500ug |
500 ÎĽg | N/A | ||||||
16015190
|
enQuireBio™
QP6696-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_065160.1 | |
Secreted Ly-6 / uPAR-related protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
57152 | |
Research Use Only | |
SLURP1 | |
Recombinant Protein | |
E. coli | |
LKCYTCKEPM TSASCRTITR CKPEDTACMT TLVTVEAEYP FNQSPVVTRS CSSSCVATDP DSIGAAHLIF CCFRDLCNSE L | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Secreted Ly-6 / uPAR-related protein 1