missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SH3GL2 Protein

A cDNA sequence encoding the SH3GL2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13490-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943739

  • 2405.00 SEK / 20µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SH3GL2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSVAGL KKQFHKATQK VSEKVGGAEG TKLDDDFKEM ERKVDVTSRA VMEIMTKTIE YLQPNPASRA KLSMINTMSK IRGQEKGPGY PQAEALLAEA MLKFGRELGD DCNFGPALGE VGEAMRELSE VKDSLDIEVK QNFIDPLQNL HDKDLREIQH HLKKLEGRRL DFDYKKKRQG KIPDEELRQA LEKFDESKEI AESSMFNLLE MDIEQVSQLS ALVQAQLEYH KQAVQILQQV TVRLEERIRQ ASSQPRREYQ PKPRMSLEFP TGDSTQPNGG LSHTGTPKPS GVQMDQPCCR ALYDFEPENE GELGFKEGDI ITLTNQIDEN WYEGMLHGHS GFFPINYVEI LVALPH
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SH3GL2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 50mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers