missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH3GL2 Protein
A cDNA sequence encoding the SH3GL2 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | SH3GL2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953739
|
enQuireBio™
QP13490-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943739
|
enQuireBio™
QP13490-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933739
|
enQuireBio™
QP13490-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SH3GL2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SH3GL2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 50mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSVAGL KKQFHKATQK VSEKVGGAEG TKLDDDFKEM ERKVDVTSRA VMEIMTKTIE YLQPNPASRA KLSMINTMSK IRGQEKGPGY PQAEALLAEA MLKFGRELGD DCNFGPALGE VGEAMRELSE VKDSLDIEVK QNFIDPLQNL HDKDLREIQH HLKKLEGRRL DFDYKKKRQG KIPDEELRQA LEKFDESKEI AESSMFNLLE MDIEQVSQLS ALVQAQLEYH KQAVQILQQV TVRLEERIRQ ASSQPRREYQ PKPRMSLEFP TGDSTQPNGG LSHTGTPKPS GVQMDQPCCR ALYDFEPENE GELGFKEGDI ITLTNQIDEN WYEGMLHGHS GFFPINYVEI LVALPH | |
Greater than 90.0% as determined by SDS-PAGE. |