missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIRPG / SIRP gamma / CD172g Protein

A DNA sequence encoding the Homo sapiens (Human) SIRPG / SIRP gamma / CD172g, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6687-ec-1mg

 View more versions of this product

Product Code. 16035110

  • 28125.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001034597.1
SIRPG / SIRP gamma / CD172g Protein
Research Use Only
SIRPG
Recombinant Protein
E. coli
EEELQMIQPE KLLLVTVGKT ATLHCTVTSL LPVGPVLWFR GVGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRISS ITPADVGTYY CVKFRKGSPE NVEFKSGPGT EMALGAKPSA PVVLGPAART TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP TGQSVAYSIR STARVVLDPW DVRSQVICEV AHVTLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRVGNQVN VTCQVRKFYP QSLQLTWSEN GNVCQRETAS TLTENKDGTY NWTSWFLVNI SDQRDDVVLT CQVKHDGQLA VSKRLALEVT VHQKDQSSDA TP
Greater than 90% as determined by SDS-PAGE.
55423
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human SIRPG / SIRP gamma / CD172g Protein > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.