Learn More
enQuireBio™ Recombinant Human SOSS complex subunit B2 Protein
A DNA sequence encoding the Homo sapiens (Human) SOSS complex subunit B2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7959-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001026886.1 | |
SOSS complex subunit B2 Protein | |
Research Use Only | |
NABP1 | |
Recombinant Protein | |
E. coli | |
MNRVNDPLIF IRDIKPGLKN LNVVFIVLEI GRVTKTKDGH EVRSCKVADK TGSITISVWD EIGGLIQPGD IIRLTRGYAS MWKGCLTLYT GRGGELQKIG EFCMVYSEVP NFSEPNPDYR GQQNKGAQSE QKNNSMNSNM GTGTFGPVGN GVHTGPESRE HQFSHAGRSN GRGLINPQLQ GTASNQTVMT TISNGRDPRR AFKR | |
Greater than 90% as determined by SDS-PAGE. |
64859 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Human SOSS complex subunit B2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction